Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272275.1 | NP_001259204.1 | 2 - 3 | 1856 | 95 | |
NM_080323.4 | NP_525062.2 | 2 - 3 | 1860 | 95 | ||
GenBank mRNA | AY058627.1 | - | 2 - 3 | 1836 | 95 | |
U77370.2 | - | 2 - 3 | 1398 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster Inventoried |