Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lva | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297920.1 | NP_001284849.1 | 7 - 8 | 8179 | 55 | |
NM_080325.4 | NP_525064.1 | 6 - 7 | 8499 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |