Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lva | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297920.1 | NP_001284849.1 | 8 - 9 | 8298 | 84 | |
NM_080325.4 | NP_525064.1 | 7 - 8 | 8618 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |