Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pp2C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297938.1 | NP_001284867.1 | 1 - 2 | 1782 | 77 | |
NM_080335.3 | NP_525074.2 | 1 - 2 | 1782 | 77 | ||
GenBank mRNA | AY069593.1 | - | 1 - 2 | 1751 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |