Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hrs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080360.5 | NP_525099.3 | 4 - 5 | 896 | 70 | |
NM_164496.1 | NP_722830.2 | 5 - 6 | 847 | 70 | ||
NM_164497.1 | NP_722831.1 | 2 - 3 | 433 | 70 | ||
GenBank mRNA | AY051789.1 | - | 4 - 5 | 880 | 70 | |
AY071846.1 | - | 4 - 5 | 845 | 70 | ||
BT053687.1 | - | 4 - 5 | 896 | 70 | ||
BT099905.1 | - | 2 - 3 | 433 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |