Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop60B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014547.4 | NP_001014547.1 | 4 - 5 | 989 | 68 | |
NM_001169815.3 | NP_001163286.1 | 4 - 5 | 989 | 68 | ||
NM_001169818.3 | NP_001163289.1 | 4 - 5 | 989 | 68 | ||
NM_080381.5 | NP_525120.1 | 4 - 5 | 989 | 68 | ||
GenBank mRNA | AF017230.1 | - | 4 - 5 | 986 | 68 | |
AF089837.1 | - | 4 - 5 | 985 | 68 | ||
DQ857345.1 | - | 4 - 5 | 988 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |