Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frq1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038764.2 | NP_001033853.1 | 3 - 4 | 669 | 126 | |
NM_001272744.1 | NP_001259673.1 | 3 - 4 | 531 | 126 | ||
NM_001272745.1 | NP_001259674.1 | 3 - 4 | 527 | 126 | ||
NM_133043.3 | NP_573271.1 | 3 - 4 | 623 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |