Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tgy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298496.1 | NP_001285425.1 | 1 - 2 | 559 | 69 | |
NM_133121.5 | NP_573349.4 | 1 - 2 | 559 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |