Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RhoGAP18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144748.2 | NP_001138220.1 | 3 - 4 | 1758 | 60 | |
NM_001144749.2 | NP_001138221.1 | 3 - 4 | 856 | 60 | ||
NM_001298497.1 | NP_001285426.1 | 3 - 4 | 954 | 60 | ||
NM_133123.2 | NP_573351.1 | 2 - 3 | 1375 | 60 | ||
NM_133124.4 | NP_573352.2 | 4 - 5 | 3383 | 60 | ||
GenBank mRNA | AY061332.1 | - | 2 - 3 | 1357 | 60 | |
DQ486141.1 | - | 3 - 4 | 838 | 60 | ||
DQ486142.1 | - | 3 - 4 | 1754 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |