Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mec2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133129.2 | NP_573357.1 | 2 - 3 | 758 | 86 | |
GenBank mRNA | AY061599.1 | - | 2 - 3 | 758 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |