Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ank | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169348.2 | NP_001162819.1 | 7 - 8 | 2768 | 91 | |
NM_175926.3 | NP_787122.1 | 6 - 7 | 2726 | 91 | ||
NM_175927.3 | NP_787123.1 | 7 - 8 | 2801 | 91 | ||
NM_175928.3 | NP_787124.1 | 7 - 8 | 2916 | 91 | ||
GenBank mRNA | BT031123.1 | - | 6 - 7 | 2721 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |