Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169404.2 | NP_001162875.1 | 2 - 3 | 1576 | 59 | |
NM_001169405.2 | NP_001162876.1 | 3 - 4 | 335 | 59 | ||
NM_001169406.2 | NP_001162877.1 | 3 - 4 | 318 | 59 | ||
NM_175970.3 | NP_787984.1 | 3 - 4 | 379 | 59 | ||
NM_175971.3 | NP_787985.1 | 2 - 3 | 351 | 59 | ||
GenBank mRNA | AY069432.1 | - | 3 - 4 | 345 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |