Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtpbeta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058030.4 | NP_477378.1 | 2 - 3 | 308 | 70 | |
GenBank mRNA | AY069124.1 | - | 2 - 3 | 265 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |