Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fpps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058032.5 | NP_477380.1 | 2 - 3 | 358 | 83 | |
GenBank mRNA | AF132554.1 | - | 2 - 3 | 357 | 83 | |
AY071445.1 | - | 2 - 3 | 353 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |