Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058093.4 | NP_477441.1 | 3 - 4 | 755 | 67 | |
GenBank mRNA | AF129084.1 | - | 3 - 4 | 704 | 67 | |
AY069760.1 | - | 3 - 4 | 748 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |