Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: drpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259620.2 | NP_001246549.1 | 4 - 5 | 1101 | 72 | |
NM_058102.3 | NP_477450.1 | 4 - 5 | 1101 | 72 | ||
GenBank mRNA | BT044170.1 | - | 4 - 5 | 1087 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |