Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gammaSnap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058121.4 | NP_477469.1 | 3 - 4 | 317 | 66 | |
GenBank mRNA | AF177988.1 | - | 3 - 4 | 247 | 66 | |
AY119512.1 | - | 3 - 4 | 267 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |