Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sur | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001347812.1 | NP_001334732.1 | 19 - 20 | 6397 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |