Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sur | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001347812.1 | NP_001334732.1 | 4 - 5 | 1324 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |