Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272236.1 | NP_001259165.1 | 5 - 6 | 1026 | 80 | |
NM_001272237.1 | NP_001259166.1 | 5 - 6 | 1008 | 80 | ||
NM_001272238.1 | NP_001259167.1 | 5 - 6 | 1023 | 80 | ||
NM_058137.6 | NP_477485.1 | 5 - 6 | 1231 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |