Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273321.1 | NP_001260250.1 | 1 - 2 | 136 | 87 | |
NM_058152.3 | NP_477500.1 | 1 - 2 | 277 | 87 | ||
GenBank mRNA | AF022891.1 | - | 1 - 2 | 439 | 87 | |
AY113208.1 | - | 1 - 2 | 277 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Drosophila melanogaster |