Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273321.1 | NP_001260250.1 | 2 - 3 | 1252 | 100 | |
NM_058152.3 | NP_477500.1 | 2 - 3 | 1393 | 100 | ||
GenBank mRNA | AF022891.1 | - | 2 - 3 | 1555 | 100 | |
AY113208.1 | - | 2 - 3 | 1393 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |