Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: beat-Ia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299046.1 | NP_001285975.1 | 4 - 5 | 1174 | 77 | |
NM_058160.4 | NP_477508.1 | 4 - 5 | 1174 | 77 | ||
GenBank mRNA | BT150361.1 | - | 4 - 5 | 1142 | 77 | |
U67057.1 | - | 4 - 5 | 1117 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |