Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs3st-B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272773.2 | NP_001259702.1 | 1 - 2 | 687 | 55 | |
NM_133142.2 | NP_573370.1 | 1 - 2 | 338 | 55 | ||
GenBank mRNA | AY119100.1 | - | 1 - 2 | 251 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |