Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKP-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272788.1 | NP_001259717.1 | 4 - 5 | 1146 | 82 | |
NM_134488.2 | NP_608332.2 | 4 - 5 | 1159 | 82 | ||
GenBank mRNA | BT031134.1 | - | 4 - 5 | 1123 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |