Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298523.1 | NP_001285452.1 | 2 - 3 | 484 | 80 | |
NM_134494.3 | NP_608338.1 | 2 - 3 | 322 | 80 | ||
GenBank mRNA | AY051833.1 | - | 2 - 3 | 303 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |