Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssu72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298526.1 | NP_001285455.1 | 1 - 2 | 317 | 65 | |
NM_134498.4 | NP_608342.1 | 1 - 2 | 317 | 65 | ||
GenBank mRNA | AY060426.1 | - | 1 - 2 | 311 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |