Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272811.1 | NP_001259740.1 | 2 - 3 | 474 | 65 | |
NM_001272812.1 | NP_001259741.1 | 2 - 3 | 479 | 65 | ||
NM_134530.3 | NP_608374.1 | 1 - 2 | 195 | 65 | ||
GenBank mRNA | AY051970.1 | - | 1 - 2 | 194 | 65 | |
BT081966.1 | - | 2 - 3 | 476 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |