Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tho2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272976.1 | NP_001259905.1 | 6 - 7 | 2082 | 102 | |
NM_134802.4 | NP_608646.3 | 6 - 7 | 2082 | 102 | ||
NM_164454.2 | NP_722763.1 | 6 - 7 | 2082 | 102 | ||
GenBank mRNA | AJ556820.2 | - | 6 - 7 | 1926 | 102 | |
BT031136.1 | - | 6 - 7 | 2070 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |