Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: toc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273060.1 | NP_001259989.1 | 1 - 2 | 249 | 64 | |
NM_164524.2 | NP_722875.2 | 1 - 2 | 249 | 64 | ||
GenBank mRNA | BT031283.1 | - | 1 - 2 | 233 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |