Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgant4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144307.2 | NP_001137779.1 | 3 - 4 | 1336 | 138 | |
NM_164539.2 | NP_722910.2 | 3 - 4 | 1468 | 138 | ||
GenBank mRNA | AF324752.1 | - | 3 - 4 | 1268 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |