Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hel25E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273171.1 | NP_001260100.1 | 2 - 3 | 392 | 54 | |
NM_164646.3 | NP_723089.1 | 2 - 3 | 392 | 54 | ||
NM_164647.1 | NP_723090.1 | 1 - 2 | 282 | 54 | ||
NM_164648.2 | NP_723091.1 | 2 - 3 | 385 | 54 | ||
GenBank mRNA | AY118921.1 | - | 2 - 3 | 347 | 54 | |
X79802.1 | - | 363 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |