Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gyc32E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103678.2 | NP_001097148.1 | 16 - 17 | 2357 | 68 | |
NM_001298905.1 | NP_001285834.1 | 15 - 16 | 2249 | 68 | ||
NM_176013.2 | NP_788027.1 | 15 - 16 | 2273 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |