Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176064.3 | NP_788078.1 | 1 - 2 | 111 | 64 | |
GenBank mRNA | AY118294.1 | - | 1 - 2 | 96 | 64 | |
DQ062758.1 | - | 1 - 2 | 80 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster Inventoried |