Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144160.2 | NP_001137632.1 | 15 - 16 | 2479 | 74 | |
NM_001273919.1 | NP_001260848.1 | 15 - 16 | 2209 | 74 | ||
NM_001273920.1 | NP_001260849.1 | 16 - 17 | 2263 | 74 | ||
NM_001273921.1 | NP_001260850.1 | 16 - 17 | 2311 | 74 | ||
NM_001273922.1 | NP_001260851.1 | 15 - 16 | 2275 | 74 | ||
NM_001273923.1 | NP_001260852.1 | 14 - 15 | 2137 | 74 | ||
NM_176119.5 | NP_788299.3 | 15 - 16 | 2218 | 74 | ||
NM_176120.5 | NP_788300.3 | 17 - 18 | 2773 | 74 | ||
GenBank mRNA | AY823300.1 | - | 15 - 16 | 2212 | 74 | |
AY823301.1 | - | 15 - 16 | 2212 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |