Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: lola | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176134.4 | NP_788314.1 | 5 - 6 | 1766 | 72 | |
NM_176135.4 | NP_788315.1 | 5 - 6 | 1760 | 72 | ||
GenBank mRNA | AB107281.1 | - | 5 - 6 | 1966 | 72 | |
AB107301.1 | - | 5 - 6 | 1907 | 72 | ||
AB107321.1 | - | 5 - 6 | 1763 | 72 | ||
AB107341.1 | - | 5 - 6 | 1763 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |