Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs3st-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299660.1 | NP_001286589.1 | 4 - 5 | 1727 | 61 | |
NM_176229.3 | NP_788409.1 | 4 - 5 | 1727 | 61 | ||
GenBank mRNA | AY121626.1 | - | 4 - 5 | 1712 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |