Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs3st-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299660.1 | NP_001286589.1 | 6 - 7 | 2358 | 79 | |
NM_176229.3 | NP_788409.1 | 6 - 7 | 2358 | 79 | ||
GenBank mRNA | AY121626.1 | - | 6 - 7 | 2343 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |