Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176240.2 | NP_788420.1 | 1 - 2 | 481 | 62 | |
GenBank mRNA | BT022158.1 | - | 1 - 2 | 453 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |