Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mthl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259603.2 | NP_001246532.1 | 3 - 4 | 1071 | 68 | |
NM_001259604.1 | NP_001246533.1 | 2 - 3 | 1369 | 68 | ||
NM_001259605.2 | NP_001246534.1 | 2 - 3 | 1369 | 68 | ||
NM_001274276.1 | NP_001261205.1 | 2 - 3 | 1291 | 68 | ||
NM_001274277.1 | NP_001261206.1 | 2 - 3 | 861 | 68 | ||
NM_138186.3 | NP_612030.2 | 2 - 3 | 861 | 68 | ||
NM_176268.3 | NP_788446.2 | 2 - 3 | 1291 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Drosophila melanogaster |