Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mthl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176295.2 | NP_788473.1 | 2 - 3 | 725 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Drosophila melanogaster |