Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SrpRbeta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274667.1 | NP_001261596.1 | 1 - 2 | 185 | 66 | |
NM_176307.3 | NP_788485.1 | 1 - 2 | 185 | 66 | ||
GenBank mRNA | AF181658.2 | - | 1 - 2 | 124 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |