Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SrpRbeta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274667.1 | NP_001261596.1 | 2 - 3 | 493 | 67 | |
NM_176307.3 | NP_788485.1 | 2 - 3 | 493 | 67 | ||
GenBank mRNA | AF181658.2 | - | 2 - 3 | 432 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |