Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176355.2 | NP_788533.1 | 3 - 4 | 496 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |