Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: janB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057236.5 | NP_476584.2 | 1 - 2 | 134 | 82 | |
GenBank mRNA | BT125584.1 | - | 1 - 2 | 134 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |