Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275407.2 | NP_001262336.1 | 4 - 5 | 2042 | 80 | |
NM_001275408.2 | NP_001262337.1 | 5 - 6 | 2480 | 80 | ||
NM_057263.5 | NP_476611.1 | 5 - 6 | 2394 | 80 | ||
NM_169177.3 | NP_731141.1 | 4 - 5 | 2160 | 80 | ||
GenBank mRNA | AY051979.1 | - | 4 - 5 | 2221 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |