Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275407.2 | NP_001262336.1 | 8 - 9 | 4162 | 61 | |
NM_001275408.2 | NP_001262337.1 | 10 - 11 | 5635 | 61 | ||
NM_057263.5 | NP_476611.1 | 9 - 10 | 4514 | 61 | ||
NM_169177.3 | NP_731141.1 | 9 - 10 | 5315 | 61 | ||
GenBank mRNA | AY051979.1 | - | 9 - 10 | 5376 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |