Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300640.1 | NP_001287569.1 | 3 - 4 | 403 | 84 | |
NM_057291.4 | NP_476639.1 | 3 - 4 | 403 | 84 | ||
NM_057292.4 | NP_476640.1 | 3 - 4 | 403 | 84 | ||
GenBank mRNA | AY070972.1 | - | 3 - 4 | 420 | 84 | |
K01567.1 | - | 3 - 4 | 381 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |