Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abd-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170161.2 | NP_001163632.1 | 2 - 3 | 1596 | 67 | |
NM_001260216.2 | NP_001247145.1 | 2 - 3 | 1893 | 67 | ||
NM_057345.5 | NP_476693.1 | 3 - 4 | 1495 | 67 | ||
NM_169733.5 | NP_732176.1 | 1 - 2 | 972 | 67 | ||
GenBank mRNA | BT016031.1 | - | 2 - 3 | 1883 | 67 | |
X54453.1 | - | 758 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |