Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptp99A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202380.2 | NP_001189309.1 | 3 - 4 | 1134 | 76 | |
NM_057468.3 | NP_476816.1 | 7 - 8 | 2073 | 76 | ||
NM_143434.5 | NP_651691.4 | 14 - 15 | 3090 | 76 | ||
NM_170409.3 | NP_733288.1 | 8 - 9 | 2204 | 76 | ||
NM_206579.3 | NP_996302.1 | 8 - 9 | 2249 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |